Please login to your online account to display your discounted pricing

Potassium hydride, 25 to 35 wt.%, dispersion in mineral oil, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC387961000

 View more versions of this product

Catalog No. AC387961000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 40.11
Physical Form Dispersion of Powder In Mineral Oil
Color Grayish beige
Quantity 100g
Density 1.5400g/mL
Packaging Plastic bottle
CAS 8042-47-5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Potassium hydride
CAS 7693-26-7
Molecular Formula HK
Linear Formula KH
Health Hazard 1 GHS Signal Word: Danger
Concentration 25.0 to 35.0 wt%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

Specifications

Chemical Name or Material Potassium hydride
CAS 7693-26-7
CAS 8042-47-5
Molecular Formula HK
Linear Formula KH
Formula Weight 40.11
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

SDS & Safety Documents