Please login to your online account to display your discounted pricing

Potassium hydride, 25 to 35 wt.%, dispersion in mineral oil, ACROS Organics™

$99.62 - $257.92

Chemical Identifiers

Chemical Name or Material Potassium Hydride
CAS 7693-26-7, 8042-47-5
CAS 8042-47-5
Molecular Formula HK
Linear Formula KH
Formula Weight 40.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC387960250

  SDS
thermo fisher scientific
AC387960250
Plastic Bottle in Tin Can 25g Each for $99.62

AC387961000

  SDS
thermo fisher scientific
AC387961000
Plastic bottle 100g Each for $257.92

AC387965000

  SDS
thermo fisher scientific
AC387965000
Plastic Drum 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Grayish beige
Formula Weight 40.11
Physical Form Dispersion of Powder In Mineral Oil
Quantity 25g
Packaging Plastic Bottle in Tin Can
Density 1.5400g/mL
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 7693-26-7
Molecular Formula HK
Chemical Name or Material Potassium hydride
CAS 8042-47-5
Linear Formula KH
Concentration 25.0 to 35.0 wt%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

Reduction reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.