Please login to your online account to display your discounted pricing

Potassium hydride, 25 to 35 wt.%, dispersion in mineral oil, ACROS Organics™


Quantity 500g
Packaging Plastic Drum

Manufacturer: thermo fisher scientific  AC387965000

 View more versions of this product

Catalog No. AC387965000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Dispersion of Powder In Mineral Oil
Formula Weight 40.11
Color Grayish beige
Quantity 500g
Chemical Name or Material Potassium Hydride
Packaging Plastic Drum
CAS 7693-26-7, 8042-47-5
UN Number 3129
Boiling Point 316.0°C
Density 1.5400g/mL
Linear Formula KH
Molecular Formula HK
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
CAS 8042-47-5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Concentration 25.0 to 35.0 wt%

Specifications

Chemical Name or Material Potassium Hydride
CAS 7693-26-7, 8042-47-5
CAS 8042-47-5
Molecular Formula HK
Linear Formula KH
Formula Weight 40.11
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

SDS & Safety Documents