missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexafluorozirconate, 99%

For manufacture of metallic zirconium

Manufacturer:  Alfa Aesar™ 8610236

 View more versions of this product

Catalog No. AA8610236



Specifications

Specifications

Potassium hexafluorozirconate
F6K2Zr
UN3288
potassium hexafluorozirconate, zirconium potassium fluoride, potassium fluorozirconate, f6zr.2k, dipotassium hexafluorozirconate, potassium hexafluorozirconate iv, dipotassium hexafluorozirconiumdiuide, dipotassium hexafluorozirconate iv
BJZIJOLEWHWTJO-UHFFFAOYSA-H
dipotassium;hexafluorozirconium(2-)
56828182
99%
−10 Mesh
500g
16923-95-8
MFCD00042547
14,7639
Slightly soluble in cold water, soluble in hot water
F[Zr-2](F)(F)(F)(F)F.[K+].[K+]
283.411
283.41
3.48g/mL
Odorless

Chemical Identifiers

16923-95-8
283.411
BJZIJOLEWHWTJO-UHFFFAOYSA-H
56828182
F[Zr-2](F)(F)(F)(F)F.[K+].[K+]
F6K2Zr
MFCD00042547
potassium hexafluorozirconate, zirconium potassium fluoride, potassium fluorozirconate, f6zr.2k, dipotassium hexafluorozirconate, potassium hexafluorozirconate iv, dipotassium hexafluorozirconiumdiuide, dipotassium hexafluorozirconate iv
dipotassium;hexafluorozirconium(2-)
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents