missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hexafluorosilicate, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 89023A1

 View more versions of this product

Catalog No. AA89023A1



Specifications

Specifications

Potassium hexafluorosilicate
99%
MFCD00040666
UUHPPUWEDCVPCO-UHFFFAOYSA-N
dipotassium;hexafluorosilicon(2-)
61851
Powder
White
16871-90-2
F6K2Si
potassium hexafluorosilicate, potassium fluosilicate, potassium silicofluoride, dipotassium hexafluorosilicate, silicate 2-, hexafluoro-, dipotassium, unii-2610w49xyd, dipotassium hexafluorosilicon 2-, dipotassium hexafluorosilicate 2-, sif6.2k, potassium hexafluorosilicate iv
F[Si-2](F)(F)(F)(F)F.[K+].[K+]
220.272
220.28
2.27g/cm3 at 20°C
1kg

Chemical Identifiers

16871-90-2
220.272
UUHPPUWEDCVPCO-UHFFFAOYSA-N
61851
F[Si-2](F)(F)(F)(F)F.[K+].[K+]
F6K2Si
MFCD00040666
potassium hexafluorosilicate, potassium fluosilicate, potassium silicofluoride, dipotassium hexafluorosilicate, silicate 2-, hexafluoro-, dipotassium, unii-2610w49xyd, dipotassium hexafluorosilicon 2-, dipotassium hexafluorosilicate 2-, sif6.2k, potassium hexafluorosilicate iv
dipotassium;hexafluorosilicon(2-)
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

H301-H311-H331: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 240-896-2

SDS Documents