missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hexafluorophosphate, 99%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 191090500

 View more versions of this product

Catalog No. AC191090500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium hexafluorophosphate
17084-13-8
Crystalline Powder
98.5% min
KPF6
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
YZDGRYDIGCWVND-UHFFFAOYSA-N
potassium;hexafluorophosphate
23688904
99%
0.1% max. (100°C)
575.0°C
White
50 g
F6KP
MFCD00011412
Solubility in water: 93g/L (25°C)
F[P-](F)(F)(F)(F)F.[K+]
184.07
184.07
Extra Pure
Plastic bottle

Chemical Identifiers

17084-13-8
184.07
YZDGRYDIGCWVND-UHFFFAOYSA-N
23688904
F[P-](F)(F)(F)(F)F.[K+]
F6KP
MFCD00011412
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
potassium;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica

GHS Signal Word: Danger

EINECSNumber : 241-143-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only