missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexafluorophosphate, 95% min

Manufacturer:  Alfa Aesar™ 1153130

 View more versions of this product

Catalog No. AA1153130



Specifications

Specifications

Potassium hexafluorophosphate
F6KP
UN3260
YZDGRYDIGCWVND-UHFFFAOYSA-N
potassium;hexafluorophosphate
23688904
Crystalline
2.55g/mL
Hygroscopic
575°C
250g
17084-13-8
MFCD00011412
kpf6
F[P-](F)(F)(F)(F)F.[K+]
184.062
184.07
≤95%
Odorless
White
3.0 to 7.0

Chemical Identifiers

17084-13-8
184.062
YZDGRYDIGCWVND-UHFFFAOYSA-N
23688904
F[P-](F)(F)(F)(F)F.[K+]
F6KP
MFCD00011412
kpf6
potassium;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents