Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexafluoronickelate(IV)

Chemical Identifiers

Chemical Name or Material Potassium hexafluoronickelate(IV)
CAS 17218-47-2
Formula Weight 250.91
MDL Number MFCD00083064
UN Number UN3288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3943904

  SDS
alfa aesar™
3943904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3943909

  SDS
alfa aesar™
3943909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3943918

  SDS
alfa aesar™
3943918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 250.91
Physical Form Powder
Health Hazard 2 GHS H Statement
H331-H350-H302-H312-H317
Toxic if inhaled.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
UN Number UN3288
CAS 17218-47-2
Quantity 2g
Health Hazard 3 GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Potassium hexafluoronickelate(IV)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
MDL Number MFCD00083064
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H350-H302-H312-H317
Toxic if inhaled.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.