missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hexafluoroarsenate(V), 99% (metals basis), Alfa aesar™

Chemical Identifiers

CAS 17029-22-0
Physical Form Powder
Assay Percent Range 99% (metals basis)
Molecular Formula KAsF6
Formula Weight 228.01
MDL Number MFCD00042546
Chemical Name or Material Potassium hexafluoroarsenate(V)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1153206 SDS Alfa Aesar
1153206
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1153214 SDS Alfa Aesar
1153214
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

17029-22-0
99% (metals basis)
228.01
Potassium hexafluoroarsenate(V)
Powder
KAsF6
MFCD00042546

Specifications

228.01
White
Acrid
White powder
KAsF6
Potassium hexafluoroarsenate(V)
5g
Powder
17029-22-0
MFCD00042546
99% (metals basis)

Safety and Handling

H301-H331-H400-H410: Toxic if swallowed. Toxic if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 241-102-7