Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexafluoroantimonate, 98% min

$72.18 - $72.18

Chemical Identifiers

Chemical Name or Material Potassium hexafluoroantimonate
CAS 16893-92-8
Formula Weight 274.84
MDL Number MFCD00043080
UN Number UN1549
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4014609

  SDS
alfa aesar™
4014609
10g Each for $72.18
Description & Specifications

Specifications

Formula Weight 274.84
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Health Hazard 1 Warning
Boiling Point 1505°C
Chemical Name or Material Potassium hexafluoroantimonate
Assay ≤98%
UN Number UN1549
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
Melting Point 846°C
MDL Number MFCD00043080
CAS 16893-92-8
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Physical Form Powder
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.