missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hexafluoroantimonate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 358740250

 View more versions of this product

Catalog No. AC358740250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Potassium hexafluoroantimonate
Crystalline Powder
KF6Sb
274.87
97.5% min.
White to light beige
16893-92-8
98%
KSbF6
MFCD00043080
Glass bottle
25g

Chemical Identifiers

98%
KSbF6
MFCD00043080
KF6Sb
274.87
Potassium hexafluoroantimonate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning