Please login to your online account to display your discounted pricing

Potassium hexafluoroantimonate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Potassium hexafluoroantimonate
Assay Percent Range 98%
Molecular Formula KF6Sb
Linear Formula KSbF6
Formula Weight 274.87
MDL Number MFCD00043080
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC358740250

  SDS
thermo fisher scientific
358740250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Quantity 25g
Color White to light beige
Formula Weight 274.87
Chemical Name or Material Potassium hexafluoroantimonate
Linear Formula KSbF6
CAS 16893-92-8
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Elemental Analysis 97.5% min.
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula KF6Sb
MDL Number MFCD00043080
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.