Learn More
Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01187609
Description
Potassium hexacyanoruthenate(II) hydrate used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium hexacyanoruthenate(II) hydrate | |
White | |
10 g | |
K4Ru(CN)6·xH2O | |
UN1588 | |
Soluble in water. | |
[K+].[K+].[K+].[K+].N#C[Ru++](C#N)(C#N)(C#N)(C#N)C#N | |
413.57 | |
413.57 (Anhydrous) |
339268-21-2 | |
Crystalline | |
C6K4N6Ru | |
MFCD00168064 | |
potassium hexacyanoruthenate ii hydrate, tetrapotassium hexacyanorutheniumtetrauide hydrate | |
FULSJSKUKUZHLZ-UHFFFAOYSA-N | |
tetrapotassium;ruthenium(2+);hexacyanide;hydrate | |
71311369 | |
Ru 23.0% min. |
Chemical Identifiers
339268-21-2 | |
413.57 | |
FULSJSKUKUZHLZ-UHFFFAOYSA-N | |
71311369 | |
[K+].[K+].[K+].[K+].N#C[Ru++](C#N)(C#N)(C#N)(C#N)C#N |
C6K4N6Ru | |
MFCD00168064 | |
potassium hexacyanoruthenate ii hydrate, tetrapotassium hexacyanorutheniumtetrauide hydrate | |
tetrapotassium;ruthenium(2+);hexacyanide;hydrate |
Safety and Handling
GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.
EINECSNumber : 239-097-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only