missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexacyanoplatinate(IV)

Manufacturer:  Alfa Aesar™ 12678MD

 View more versions of this product

Catalog No. AA12678MD



Specifications

Specifications

Potassium hexacyanoplatinate(IV)
C6K2N6Pt
MFCD00058745
dipotassium hexacyanoplatinumdiuide
YHKPKKPXZORMRU-UHFFFAOYSA-N
dipotassium;platinum(4+);hexacyanide
22597308
Powder
White
16920-94-8
K2Pt(CN)6
UN1588
no data available
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[K+].[K+].[Pt+4]
429.389
429.4
Odorless
250mg

Chemical Identifiers

16920-94-8
429.389
YHKPKKPXZORMRU-UHFFFAOYSA-N
22597308
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[K+].[K+].[Pt+4]
C6K2N6Pt
MFCD00058745
dipotassium hexacyanoplatinumdiuide
dipotassium;platinum(4+);hexacyanide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.

EINECSNumber : 240-978-8

TSCA : No

Recommended Storage : Ambient temperatures

Documents