missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hexacyanoferrate(III), ACS, 99.0% min.; Alfa Aesar™

$110.09 - $110.09

Chemical Identifiers

CAS 13746-66-2
Physical Form Crystalline
Assay Percent Range 99.0% min.
Molecular Formula K3Fe(CN)6
Formula Weight 329.26
MDL Number MFCD00011392
Synonym Potassium ferricyanide
Chemical Name or Material Potassium hexacyanoferrate(III)
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3335736 SDS Alfa Aesar
3335736
500g Each for $110.09
AA33357A3 SDS Alfa Aesar
33357A3
2kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

13746-66-2
99.0% min.
329.26
Potassium ferricyanide
ACS Grade
Crystalline
K3Fe(CN)6
MFCD00011392
Potassium hexacyanoferrate(III)

Specifications

13746-66-2
99.0% min.
Orange crystalline
MFCD00011392
1.89g/cm3 at 20°C
Light sensitive
500g
ACS Grade
Crystalline
K3Fe(CN)6
329.26
Potassium ferricyanide
Maximum level of impurities: Insoluble matter 0.005%, Cl 0.01%, SO4 0.01%, Ferro compounds 0.05% as ferrocyanide radical [Fe(CN)6]4-
Orange
Potassium hexacyanoferrate(III)

Safety and Handling

This material and/or its container must be disposed of as hazardous waste.

H303: May be harmful if swallowed.

EINECSNumber : 237-323-3