Please login to your online account to display your discounted pricing

Potassium hexacyanoferrate(II) trihydrate, ACS, 98.5-102.0%, Alfa Aesar™

$50.62 - $50.62

Chemical Identifiers

Chemical Name or Material Potassium hexacyanoferrate(II) trihydrate
CAS 14459-95-1
Purity Grade ACS Grade
Molecular Formula K4Fe(CN)6·3H2O
Formula Weight 422.39 (368.36 Anhydrous)
MDL Number MFCD00167023
Merck Index 14,7631
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3335822

  SDS
alfa aesar
3335822
Each for $50.62
Description & Specifications

Specifications

Formula Weight 422.39 (368.36 Anhydrous)
Purity Grade ACS Grade
Melting Point 69° to 71°C
Quantity 100g
Odor Odorless
MDL Number MFCD00167023
Physical Form Crystalline
Merck Index 14,7631
Color Pale Yellow
Molecular Formula K4Fe(CN)6·3H2O
Assay 98.5 to 102.0%
TSCA TSCA
Density 1.85
pH 9.5
Chemical Name or Material Potassium hexacyanoferrate(II) trihydrate
CAS 14459-95-1