Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexacyanoferrate(II), 0.1N Standardized Solution

Chemical Identifiers

Chemical Name or Material Potassium hexacyanoferrate(II)
CAS 13943-58-3
Name Note Standardized Solution
Linear Formula K4Fe(CN)6
Formula Weight 368.36
MDL Number MFCD00167022
Merck Index 14,7631
Synonym Potassium ferrocyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35585K2

  SDS
alfa aesar™
35585K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 368.36
Concentration 0.1N
Merck Index 14,7631
Chemical Name or Material Potassium hexacyanoferrate(II)
Quantity 1L
Physical Form Liquid
TSCA TSCA
Name Note Standardized Solution
Linear Formula K4Fe(CN)6
Odor Odorless
Color Pale yellow
CAS 13943-58-3
MDL Number MFCD00167022
Synonym Potassium ferrocyanide