missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexacyanocobaltate(III), 90+%

Manufacturer:  Alfa Aesar™ 2312618

 View more versions of this product

Catalog No. AA2312618



Specifications

Specifications

Potassium hexacyanocobaltate(III)
K3[Co(CN)6]
14,7637
332.33
≥90%
Odorless
50g
13963-58-1
MFCD00011391
Soluble in water and acetic acid solutions; Insoluble in alcohol
Crystalline
1.906g/mL
Brown to Yellow

Chemical Identifiers

13963-58-1
MFCD00011391
Safety and Handling

Safety and Handling

GHS H Statement
H311-H351-H302-H332-H317
Toxic in contact with skin.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : GF9470000

TSCA : TSCA

SDS Documents