Please login to your online account to display your discounted pricing

Potassium hexachlororuthenate(IV), 99.95% (Metal basis), Ru 25.3% min., Alfa Aesar™, Premion™

Chemical Identifiers

Chemical Name or Material Potassium hexachlororuthenate(IV)
CAS 23013-82-3
Physical Form Crystalline Powder
Assay Percent Range 99.95% (metals basis)
Molecular Formula K2RuCl6
Formula Weight 391.99
MDL Number MFCD00050159
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4039903

 
alfa aesar
4039903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4039906

 
alfa aesar
4039906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 391.99
Quantity 1g
Color Red-brown
Chemical Name or Material Potassium hexachlororuthenate(IV)
EINECS Number 245-381-6
Assay Percent Range 99.95% (metals basis)
Molecular Formula K2RuCl6
Appearance Red-browm crystalline powder
MDL Number MFCD00050159
CAS 23013-82-3
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Composition Ru 25.3% min
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.