missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium hexachlororuthenate(IV), Premion™, 99.95% (metal basis), Ru 25.3% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04039906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium hexachlororuthenate(IV) | |
23013-82-3 | |
5 g | |
K2RuCl6 | |
391.99 |
Red to Brown | |
Crystalline Powder | |
99.95% (metals basis) | |
MFCD00050159 | |
Red-browm crystalline powder |
Chemical Identifiers
23013-82-3 | |
K2RuCl6 | |
MFCD00050159 |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 245-381-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only