Learn More
Potassium hexachloroplatinate(IV), ca. 40% Pt, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 195350250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Potassium hexachloroplatinate(IV) | |
25g | |
K2PtCl6 | |
potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride | |
DPAIVKJGTXERIM-UHFFFAOYSA-H | |
dipotassium;hexachloroplatinum(2-) | |
61856 | |
ca. 40% Pt |
16921-30-5 | |
Cl6K2Pt | |
15,7757 | |
Solubility in water: 50g/L (95°C). Other solubilities: insoluble in alcohol | |
Cl[Pt-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+] | |
486.01 | |
486.01 | |
Glass bottle |
Safety and Handling
GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
May be corrosive to metals.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
GHS Signal Word: Danger
EINECSNumber : 240-979-3
RUO â Research Use Only