Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexachloropalladate(IV), Pd 26.3% min

Chemical Identifiers

Chemical Name or Material Potassium hexachloropalladate(IV)
CAS 16919-73-6
Formula Weight 397.32
MDL Number MFCD00043081
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1188403

  SDS
alfa aesar™
1188403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1188406

  SDS
alfa aesar™
1188406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1188414

  SDS
alfa aesar™
1188414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 397.32
UN Number UN3077
Odor Odorless
Quantity 1g
Physical Form Crystalline
MDL Number MFCD00043081
CAS 16919-73-6
Health Hazard 2 GHS H Statement
H318-H302-H317-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P310-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Color Red
Concentration Pd 26.3% min.
RTECS Number RT3353000
Health Hazard 1 Danger
TSCA TSCA
Sensitivity Hygroscopic
Density 2.738g/mL
Chemical Name or Material Potassium hexachloropalladate(IV)
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P310-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302-H317-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes skin irritation.