missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexachloroiridate(IV), Ir 39% min

Manufacturer:  Alfa Aesar™ 1188806

 View more versions of this product

Catalog No. AA1188806



Specifications

Specifications

Potassium hexachloroiridate(IV)
Cl6IrK2
cl6ir.2k, dipotassium hexachloroiridate, dipotassium hexachloroiridate 2-, potassium hexachloroiridate iv
WEKZJZDULNNSCD-UHFFFAOYSA-H
dipotassium;hexachloroiridium(2-)
11049163
Crystalline
Odorless
(decomposition)
16920-56-2
MFCD00011369
Insoluble in water.
Cl[Ir-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+]
483.114
483.12
Ir 39% min.
Hygroscopic
5g

Chemical Identifiers

16920-56-2
483.114
WEKZJZDULNNSCD-UHFFFAOYSA-H
11049163
Cl[Ir-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+]
Cl6IrK2
MFCD00011369
cl6ir.2k, dipotassium hexachloroiridate, dipotassium hexachloroiridate 2-, potassium hexachloroiridate iv
dipotassium;hexachloroiridium(2-)
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 240-976-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents