missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexachloroiridate(III) hydrate, Ir 33.4% min

Manufacturer:  Alfa Aesar™ 1188703

 View more versions of this product

Catalog No. AA1188703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium hexachloroiridate(III) hydrate
MFCD00038699
Crystalline
1g
35705-96-5
522.22 (Anhydrous)
Ir 33.4% min.

Chemical Identifiers

35705-96-5
MFCD00038699
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents