Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexachloroiridate(III) hydrate, Ir 33.4% min

Chemical Identifiers

Chemical Name or Material Potassium hexachloroiridate(III) hydrate
Formula Weight 522.22 (Anhydrous)
MDL Number MFCD00038699
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1188703

  SDS
alfa aesar™
1188703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1188706

  SDS
alfa aesar™
1188706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1188714

  SDS
alfa aesar™
1188714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 522.22 (Anhydrous)
Quantity 1g
MDL Number MFCD00038699
Physical Form Crystalline
Concentration Ir 33.4% min.
Chemical Name or Material Potassium hexachloroiridate(III) hydrate
TSCA TSCA