missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexabromotellurate(IV), 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4408106

 View more versions of this product

Catalog No. AA4408106



Specifications

Specifications

Potassium hexabromotellurate (IV)
(metals basis)
MFCD03427026
potassium hexabromotellurate iv
[K+].[K+].[Br-].[Br-].Br[Te](Br)(Br)Br
121233754
Crystalline
5g
16986-18-8
Br6K2Te
UN3284
HMFRPYOIJYYQFO-UHFFFAOYSA-L
685.221
685.22
99.99%

Chemical Identifiers

16986-18-8
685.221
HMFRPYOIJYYQFO-UHFFFAOYSA-L
121233754
Br6K2Te
MFCD03427026
potassium hexabromotellurate iv
[K+].[K+].[Br-].[Br-].Br[Te](Br)(Br)Br
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TELLURIUM COMPOUND, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents