missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexabromopalladate(IV), Premion™, 99.999% (metals basis), Pd 15.5% min

Manufacturer:  Alfa Aesar™ 4394706

 View more versions of this product

Catalog No. AA4394706



Specifications

Specifications

Potassium hexabromopalladate (IV)
(metals basis)
MFCD03093651
SKCNWISEAGYZFC-UHFFFAOYSA-H
dipotassium;tetrabromopalladium;dibromide
121233751
Crystalline
Premion™
5g
16919-74-7
Br6K2Pd
potassium hexabromopalladate iv , premion
[K+].[K+].[Br-].[Br-].Br[Pd](Br)(Br)Br
664.041
664.04
99.999%
Pd 15.5% min

Chemical Identifiers

16919-74-7
664.041
SKCNWISEAGYZFC-UHFFFAOYSA-H
121233751
[K+].[K+].[Br-].[Br-].Br[Pd](Br)(Br)Br
Br6K2Pd
MFCD03093651
potassium hexabromopalladate iv , premion
dipotassium;tetrabromopalladium;dibromide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents