Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium hexabromoiridate(IV), Ir 25.5% min

Chemical Identifiers

Chemical Name or Material Potassium hexabromoiridate(IV)
CAS 19121-78-9
Formula Weight 749.86
MDL Number MFCD00054221
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1265103

  SDS
alfa aesar™
1265103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1265106

  SDS
alfa aesar™
1265106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 749.86
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310-P301+P312-P406-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant/... container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Potassium hexabromoiridate(IV)
Odor Odorless
Physical Form Crystalline
Sensitivity Hygroscopic
MDL Number MFCD00054221
CAS 19121-78-9
Color Black
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H290-H302
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
Concentration Ir 25.5% min.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310-P301+P312-P406-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant/... container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H290-H302
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.