missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium formate, 99% (water <2%), Alfa Aesar™

Manufacturer:  Alfa Aesar A145510I

 View more versions of this product

Catalog No. AAA145510I



Description

Description

Applications
Potassium formate acts as an intermediate in the formate potash process for the production of potassium. It is a deicing salt for used on roads. It serves as a corrosion inhibitor, an anti-scaling agent, a functional fluid, a processing aid and used in petroleum production.

Solubility
Soluble in water and alcohol. Insoluble in ether.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Potassium formate
5000g
99%
CHKO2
3623272
potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
WFIZEGIEIOHZCP-UHFFFAOYSA-M
potassium;formate
2735122
84.12
165°C to 168°C
590-29-4
(water <2%)
MFCD00013100
14,7633
Soluble in water
C(=O)[O-].[K+]
84.115
CHEBI:63316
99%

Chemical Identifiers

590-29-4
84.115
WFIZEGIEIOHZCP-UHFFFAOYSA-M
2735122
potassium;formate
CHKO2
MFCD00013100
potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
CHEBI:63316
C(=O)[O-].[K+]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : LQ9625000

TSCA : TSCA

storageNote1 : Hygroscopic

Documents