Please login to your online account to display your discounted pricing

Potassium Fluoride, Reagent Grade, LabChem™

$174.09 - $174.09

Chemical Identifiers

Chemical Name or Material Potassium Fluoride
CAS 7789-23-3
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula KF
Formula Weight 58.1
UN Number UN1812
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC190901

 
lab chem
LC190901
500g Each for $174.09
Description & Specifications

Specifications

Formula Weight 58.1
Packaging Poly Bottle
Linear Formula KF
CAS 7789-23-3
Boiling Point 1,505°C
Color White
Health Hazard 1 Danger
Density 2,480kg/m3
Chemical Name or Material Potassium Fluoride
EINECS Number 232-151-5
Identification Passes Test
Purity Grade Reagent Grade
Physical Form Powder
Solubility Soluble in water
Specific Gravity 2480kg/m3
Storage Information Room Temperature
UN Number UN1812
Melting Point 858°C
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Toxic if swallowed.
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Safety and Handling

Danger

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic if swallowed.