missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Fluoride, For Dissolved Oxygen, Certified, 40% (w/v), LabChem™
Supplier: LabChem LC191002
Specifications
Clear, colorless | |
FK | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
[F-].[K+] | |
1 L | |
Room Temperature |
Liquid | |
MFCD00011398 | |
Poly Bottle | |
potassium fluoride | |
58.10 | |
Potassium Fluoride |