missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium fluoride dihydrate, Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 1084118

 View more versions of this product

Catalog No. AA1084118



Specifications

Specifications

Potassium fluoride dihydrate
(metals basis)
MFCD00149929
14,7632
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium;fluoride;dihydrate
23708904
Powder
Puratronic™
Odorless
Hygroscopic
50g
13455-21-5
FH4KO2
UN1812
acmc-1byyf, ksc497c4l, potassium dihydrate fluoride, potassium fluoride dihydrate, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride kf , dihydrate, potassium fluoride, dihydrate, potassiumfluoridedihydrate, unii-95r1d12heh
O.O.[F-].[K+]
94.127
94.13 (58.10 Anhydrous)
99.995%
2.5 g/mL
1.352
41°C

Chemical Identifiers

13455-21-5
94.127
MZFXHSQFQLKKFD-UHFFFAOYSA-M
23708904
O.O.[F-].[K+]
FH4KO2
MFCD00149929
acmc-1byyf, ksc497c4l, potassium dihydrate fluoride, potassium fluoride dihydrate, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride kf , dihydrate, potassium fluoride, dihydrate, potassiumfluoridedihydrate, unii-95r1d12heh
potassium;fluoride;dihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c

H301+H311+H331-H318

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: POTASSIUM FLUORIDE, SOLID

EINECSNumber : 232-151-5

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents