Please login to your online account to display your discounted pricing

Potassium Fluoride Dihydrate, MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  15633980

 View more versions of this product

Catalog No. ICN15633980

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 94.13
Quantity 100g
Molecular Formula KF · 2H2O
CAS 13455-21-5
Recommended Storage Room Temperature
Chemical Name or Material Potassium Fluoride Dihydrate

Specifications

Chemical Name or Material Potassium Fluoride Dihydrate
CAS 13455-21-5
Molecular Formula KF · 2H2O
Formula Weight 94.13
Accessories

Room Temperature