Please login to your online account to display your discounted pricing

Potassium Fluoride Dihydrate, MP Biomedicals

$56.51 - $56.51

Chemical Identifiers

Chemical Name or Material Potassium Fluoride Dihydrate
CAS 13455-21-5
Molecular Formula KF · 2H2O
Formula Weight 94.13
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15633980

 
mp biomedicals
15633980
100g Each for $56.51
Description & Specifications

Specifications

Formula Weight 94.13
Quantity 100g
Molecular Formula KF · 2H2O
CAS 13455-21-5
Recommended Storage Room Temperature
Chemical Name or Material Potassium Fluoride Dihydrate
Safety and Handling

Room Temperature