Please login to your online account to display your discounted pricing

Potassium Fluoride AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Potassium Fluoride
Name Note Dihydrate
CAS 13455-21-5
Physical Form Crystalline
Molecular Formula KF·2H2O
Formula Weight 94.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK6944212

 
avantor performance
694406
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK6944500

 
avantor performance
694404**
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Potassium Fluoride
Name Note Dihydrate
CAS 13455-21-5
Physical Form Crystalline
Molecular Formula KF·2H2O
Formula Weight 94.13
Packaging Poly
Quantity 2.5kg