missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Fluoride AR Crystal, Macron Fine Chemicals™

macron

Manufacturer:  Avantor Performance 694404**

 View more versions of this product

Catalog No. MK6944500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium Fluoride
13455-21-5
potassium fluoride dihydrate spray dried reagentplus r, potassium fluoride dihydrate reagent grade, potassiumfluoridedihydratecrystalreagent, potassium fluoride dihydrate 250g, potassium fluoride dihydrate reagent g&, potassium fluoride dihydrate puratronic, potassium fluoride dihydrate p.a, potassium fluoride dihydrate 98.5+%, potassium fluoride kf dihydrate, potassium ion dihydrate fluoride
O.O.[F-].[K+]
94.127
94.13
POLYSTORMOR
Dihydrate
FH4KO2
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium;fluoride;dihydrate
23708904
Crystalline
500g

Chemical Identifiers

13455-21-5
94.127
potassium fluoride dihydrate spray dried reagentplus r, potassium fluoride dihydrate reagent grade, potassiumfluoridedihydratecrystalreagent, potassium fluoride dihydrate 250g, potassium fluoride dihydrate reagent g&, potassium fluoride dihydrate puratronic, potassium fluoride dihydrate p.a, potassium fluoride dihydrate 98.5+%, potassium fluoride kf dihydrate, potassium ion dihydrate fluoride
potassium;fluoride;dihydrate
FH4KO2
MZFXHSQFQLKKFD-UHFFFAOYSA-M
23708904
O.O.[F-].[K+]
Documents