Please login to your online account to display your discounted pricing

Potassium Fluoride, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$397.86 - $1,418.59

Chemical Identifiers

Chemical Name or Material Potassium Fluoride
CAS 7789-23-3
Name Note Anhydrous
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula KF
Formula Weight 58.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003760

 
avantor performance
312301
Poly Bottle 500g Each for $397.86

02003762

 
avantor performance
312305
Poly Bottle 2.5kg Each for $1,418.59
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 58.1
Chemical Name or Material Potassium Fluoride
Name Note Anhydrous
Molecular Formula KF
CAS 7789-23-3
Quantity 500g
Packaging Poly Bottle
Specific Gravity 1L = 2.48kg