Please login to your online account to display your discounted pricing

Potassium Fluoride, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Poly Bottle

Manufacturer: avantor performance  312304

 View more versions of this product

Catalog No. 02003761

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 58.1
Chemical Name or Material Potassium Fluoride
Packaging Poly Bottle
Specific Gravity 1L = 2.48kg
Molecular Formula KF
CAS 7789-23-3
Quantity 125g
Name Note Anhydrous

Specifications

Chemical Name or Material Potassium Fluoride
CAS 7789-23-3
Name Note Anhydrous
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula KF
Formula Weight 58.1