missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium fluoride, anhydrous, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 14130A1

 View more versions of this product

Catalog No. AA14130A1



Specifications

Specifications

Potassium fluoride, anhydrous
99%
MFCD00011398
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
[F-].[K+]
58.097
CHEBI:66872
Various Forms (Powder/Flake/Crystalline/Beads, Etc.)
0.13 hPa at 752°C
1kg
7789-23-3
FK
White various forms (powder/flake/crystalline/beads, etc.)
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium;fluoride
522689
58.1
Odorless
White

Chemical Identifiers

7789-23-3
58.097
NROKBHXJSPEDAR-UHFFFAOYSA-M
522689
potassium;fluoride
FK
MFCD00011398
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
CHEBI:66872
[F-].[K+]
Safety and Handling

Safety and Handling

H301-H311-H331: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled.

DOTInformation : Hazard Class: 6.1, Packing Group: III

EINECSNumber : 232-151-5

SDS Documents