Please login to your online account to display your discounted pricing

Potassium fluoride, 99%, extra pure, anhydrous, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  201350250

 View more versions of this product

Catalog No. AC201350250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder, Crystals, and/or Chunks
Purity Grade Extra Pure
Formula Weight 58.09
Quantity 25g
Color White
Merck Index 15, 7753
Assay ≥98.5%
TSCA TSCA
RTECS Number TT0700000
EINECS Number 232-151-5
Density 2.48
Fieser 01,933; 02,346; 05,555; 06,481; 08,410; 10,325; 11,434; 12,406; 13,256; 15,167; 16,282
Chloride ≤500ppm
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Assay Percent Range 99%
Linear Formula KF
Packaging Plastic bottle
MDL Number MFCD00011398
Chlorine (Cl) 500ppm max.
Iron (Fe) 50ppm max.
Sulfate ≤500ppm
Identification Pass Test
CAS 7789-23-3
Chemical Name or Material Potassium fluoride
Molecular Formula FK
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.

Specifications

Chemical Name or Material Potassium fluoride
Name Note 99%
CAS 7789-23-3
Purity Grade Extra Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula FK
Linear Formula KF
Formula Weight 58.09
MDL Number MFCD00011398
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.

SDS & Safety Documents