missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium fluoride, 99%, extra pure, anhydrous, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 201355000

 View more versions of this product

Catalog No. AC201355000



Description

Description

C-X bond formation reagent
Specifications

Specifications

Potassium fluoride
7789-23-3
FK
MFCD00011398
15, 7753
Solubility in water: 920g/L (18°C). Other solubilities: soluble in hydrofluoric acid,insoluble in alcohol unless water is present
[F-].[K+]
58.09
CHEBI:66872
Crystalline Powder, Crystals, and/or Chunks
Extra Pure
500ppm max.
Pass Test
Plastic bottle
1000ppm max
White
500g
99%
99%
KF
01,933; 02,346; 05,555; 06,481; 08,410; 10,325; 11,434; 12,406; 13,256; 15,167; 16,282
ccris 1568, fluorure de potassium, potassium fluoride, potassium fluoride kf, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, potassium fluorure, potassium fluorure french, potassiumfluoride, unii-9082wg1g3f
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium;fluoride
522689
58.09
99%
500ppm max
2.48
50ppm max.
Solubility in water: 920g/L (18°C). Other solubilities: soluble in hydrofluoric acid, insoluble in alcohol unless water is present
1505.0°C
860.0°C

Chemical Identifiers

7789-23-3
58.09
NROKBHXJSPEDAR-UHFFFAOYSA-M
522689
potassium;fluoride
FK
MFCD00011398
ccris 1568, fluorure de potassium, potassium fluoride, potassium fluoride kf, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, potassium fluorure, potassium fluorure french, potassiumfluoride, unii-9082wg1g3f
CHEBI:66872
[F-].[K+]
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air

GHS Signal Word: Danger

EINECSNumber : 232-151-5

RTECSNumber : TT0700000

TSCA : TSCA

SDS Documents