Learn More
Potassium fluoride, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 201352500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Potassium fluoride | |
7789-23-3 | |
Crystalline Powder, Crystals and/or Chunks | |
99% | |
KF | |
01,933; 02,346; 05,555; 06,481; 08,410; 10,325; 11,434; 12,406; 13,256; 15,167; 16,282 | |
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
[F-].[K+] | |
58.10 | |
CHEBI:66872 | |
≥98.5% | |
Pass Test | |
2.48 |
99% | |
White | |
250 g | |
FK | |
MFCD00011398 | |
15, 7753 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
potassium fluoride | |
522689 | |
58.09 | |
Extra Pure | |
Plastic bottle |
Chemical Identifiers
7789-23-3 | |
58.10 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
522689 | |
[F-].[K+] |
FK | |
MFCD00011398 | |
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
CHEBI:66872 |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 232-151-5
RTECSNumber : TT0700000
TSCA : TSCA
RUO – Research Use Only