Please login to your online account to display your discounted pricing

Potassium Ferrocyanide, Trihydrate, ACS Grade, 98.5 to 102.0%, LabChem™

$173.97 - $173.97

Chemical Identifiers

Chemical Name or Material Potassium Ferrocyanide
CAS 14459-95-1
CAS Max % 100
Name Note Trihydrate
Purity Grade ACS Grade
Linear Formula K4Fe(CN)6·3H2O
Formula Weight 422.39
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC190551

 
lab chem
LC190551
500g Each for $173.97
Description & Specifications

Specifications

Formula Weight 422.39
Chemical Name or Material Potassium Ferrocyanide
Identification Passes Test
Name Note Trihydrate
Packaging Amber Glass
Purity Grade ACS Grade
Physical Form Crystals
Assay 98.5 to 102.0%
Solubility Soluble in water
Specific Gravity 1.85g/cm3
Storage Information Room Temperature
Linear Formula K4Fe(CN)6·3H2O
CAS 14459-95-1
Decomposition Information Potassium oxide; Carbon monoxide; Carbon dioxide
Color Yellow
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Density 1.85g/cm3
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.