missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Ferrocyanide ACS AR Crystal, Macron Fine Chemicals™

macron

$731.50 - $731.50

Chemical Identifiers

CAS 14459-95-1
Physical Form Crystalline
Molecular Formula K4Fe(CN)6·3H2O
Formula Weight 422.39
Chemical Name or Material Potassium Ferrocyanide, Trihydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK6932212 Avantor Performance
693205
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK6932500 Avantor Performance
693204
POLYSTORMOR 500g Each for $731.50
Case of 4 Each for $2,926.00
Specifications Description & Specifications

Chemical Identifiers

14459-95-1
K4Fe(CN)6·3H2O
Potassium Ferrocyanide, Trihydrate
Crystalline
422.39
ACS Grade

Specifications

14459-95-1
K4Fe(CN)6·3H2O
Poly
Potassium Ferrocyanide, Trihydrate
Crystalline
422.39
2.5kg
ACS Grade