missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Ferricyanide, ACS Grade, ≥ 99.0%, LabChem™

$183.03 - $183.03

Chemical Identifiers

CAS 13746-66-2
CAS Max % 100
Linear Formula K3Fe(CN)6
Formula Weight 329.26
Chemical Name or Material Potassium Ferricyanide
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC190401 Lab Chem
LC190401
500g Each for $183.03
Specifications Description & Specifications

Chemical Identifiers

13746-66-2
K3Fe(CN)6
Potassium Ferricyanide
100
329.26
ACS Grade

Specifications

13746-66-2
Crystals
329.26
Passes Test
Soluble in water
Room Temperature
Orange
Potassium Ferricyanide
ACS Grade
100
K3Fe(CN)6
1.85g/cm3
Poly Bottle
1.85g/cm3
Potassium oxide; Carbon monoxide; Carbon dioxide
500g
≥99.0%

Safety and Handling

GHS H Statement
Product is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.