Please login to your online account to display your discounted pricing

Potassium ferricyanide, 98%, pure, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC196780010

 View more versions of this product

Catalog No. AC196780010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder or Crystals
Formula Weight 329.26
Quantity 1kg
Color Orange to red
Merck Index 15, 7751
EINECS Number 237-323-3
TSCA TSCA
Purity Grade Pure
Density 1.85
RTECS Number LJ8225000
Fieser 01,929; 02,345; 04,406; 05,554; 06,480; 07,300; 08,410; 09,385; 13,255
Chemical Name or Material Potassium ferricyanide
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6FeK3N6
Linear Formula K3Fe(CN)6
MDL Number MFCD00011392
CAS 13746-66-2
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Contact with acids liberates very toxic gas.
Health Hazard 3 GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Synonym Potassium hexacyanoferrate (III)
Health Hazard 1 Warning

Specifications

Chemical Name or Material Potassium ferricyanide
Name Note 98%
CAS 13746-66-2
Purity Grade Pure
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6FeK3N6
Linear Formula K3Fe(CN)6
Formula Weight 329.26
MDL Number MFCD00011392
Synonym Potassium hexacyanoferrate (III)
Accessories

Warning

GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Contact with acids liberates very toxic gas.

SDS & Safety Documents