missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium ethyl xanthate, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A114500E

 View more versions of this product

Catalog No. AAA114500E



Description

Description

Potassium ethyl xanthate is used as a reagent for extraction of metal ions into organic solvents and in the preparation of 2-mercapto benzimidazoles and 2-mercapto benzoxazoles by reacting with o-phenylenediamines and o-amino phenols respectively. It acts as a precursor in the production of unsymmetrical sulfides through cross-coupling reaction and as a floatation agent in the mining industry. It is involved in the determination of cadmium in water samples by substoichiometric radiochemical method and the adsorption of colloidal dye disperse Blue 3 onto pretreated polyester fabric.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium ethyl xanthate is used as a reagent for extraction of metal ions into organic solvents and in the preparation of 2-mercapto benzimidazoles and 2-mercapto benzoxazoles by reacting with o-phenylenediamines and o-amino phenols respectively. It acts as a precursor in the production of unsymmetrical sulfides through cross-coupling reaction and as a floatation agent in the mining industry. It is involved in the determination of cadmium in water samples by substoichiometric radiochemical method and the adsorption of colloidal dye disperse Blue 3 onto pretreated polyester fabric.

Solubility
Soluble in water.

Notes
Light sensitive. Incompatible with strong acids, strong bases and strong oxidizing agents and water.
Specifications

Specifications

Potassium ethyl xanthate
∼205°C (decomposition)
9 to 10
98%
C3H5KOS2
2500 g
14,7696
Soluble in water.
[K+].CCOC([S-])=S
160.29
160.3
140-89-6
1.558
205°C (401°F)
Unpleasant
MFCD00004931
3596974
potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide
JCBJVAJGLKENNC-UHFFFAOYSA-M
potassium;ethoxymethanedithioate
2735045
98%

Chemical Identifiers

140-89-6
160.29
JCBJVAJGLKENNC-UHFFFAOYSA-M
2735045
[K+].CCOC([S-])=S
C3H5KOS2
MFCD00004931
potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide
potassium;ethoxymethanedithioate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 205-439-3

RTECSNumber : FG1575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only