Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium ethoxide, 99+% (metals basis), 15% w/v in ethanol

Chemical Identifiers

Chemical Name or Material Potassium ethoxide
CAS 917-58-8
Linear Formula KOCH2CH3
Formula Weight 84.16
MDL Number MFCD00054277
UN Number UN3274
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39161AC

 
alfa aesar™
39161AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39161AE

 
alfa aesar™
39161AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 84.16
Chemical Name or Material Potassium ethoxide
TSCA TSCA
Physical Form Liquid
Assay ≥99%
Assay Percent Notes (metals basis)
Linear Formula KOCH2CH3
MDL Number MFCD00054277
CAS 917-58-8
UN Number UN3274
Odor Alcohol-like
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Colorless to pale yellow
Concentration 15% w/v in ethanol
Quantity 25mL
Sensitivity Air & Moisture Sensitive
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.