missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium ethoxide, 95%

Manufacturer:  Alfa Aesar™ 1426330

 View more versions of this product

Catalog No. AA1426330



Specifications

Specifications

Potassium ethoxide
Air & Moisture Sensitive
250°C (decomposition)
917-58-8
KOCH2CH3
UN3206
Soluble in ethanol, ether
CC[O-].[K+]
84.159
84.16
95%
Alcohol-like
Brown to Yellow
250g
C2H5KO
MFCD00054277
potassium ethoxide, potassium ethanolate, potassium ethylate, ethanol, potassium salt 1:1, potassium ethoxide solution, ethanol, potassium salt, potassiumethoxide, potassium alcohol, etok, koet
RPDAUEIUDPHABB-UHFFFAOYSA-N
potassium;ethanolate
23670592
Powder

Chemical Identifiers

917-58-8
84.159
RPDAUEIUDPHABB-UHFFFAOYSA-N
23670592
CC[O-].[K+]
C2H5KO
MFCD00054277
potassium ethoxide, potassium ethanolate, potassium ethylate, ethanol, potassium salt 1:1, potassium ethoxide solution, ethanol, potassium salt, potassiumethoxide, potassium alcohol, etok, koet
potassium;ethanolate
Safety and Handling

Safety and Handling

GHS H Statement
H251-H314
Self-heating: may catch fire.
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.

TSCA : TSCA

Documents