missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium diphosphate, 98%, extra pure, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC269230025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Potassium diphosphate | |
7320-34-5 | |
1090°C | |
98% | |
K4O7P2 | |
15, 7783 | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol | |
[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O | |
330.33 | |
330.34 | |
98% | |
10ppm max. | |
Plastic bottle | |
100ppm max |
Extra Pure | |
White | |
2.5kg | |
K4O7P2 | |
MFCD00011393 | |
diphosphoric acid, tetrapotassium salt, potassium diphosphate, potassium pyrophosphate, pyrophosphoric acid, tetrapotassium salt, tetrapotassium diphosphate, tetrapotassium diphosphorate, tetrapotassium phosphonato phosphate, tetrapotassium pyrophosphate, tkpp, unii-b9w4019h5g | |
RYCLIXPGLDDLTM-UHFFFAOYSA-J | |
tetrapotassium;phosphonato phosphate | |
23740 | |
Crystalline Powder | |
Extra Pure | |
10ppm max. | |
20ppm max. | |
50ppm max. |
Chemical Identifiers
7320-34-5 | |
330.33 | |
RYCLIXPGLDDLTM-UHFFFAOYSA-J | |
23740 | |
[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O |
K4O7P2 | |
MFCD00011393 | |
diphosphoric acid, tetrapotassium salt, potassium diphosphate, potassium pyrophosphate, pyrophosphoric acid, tetrapotassium salt, tetrapotassium diphosphate, tetrapotassium diphosphorate, tetrapotassium phosphonato phosphate, tetrapotassium pyrophosphate, tkpp, unii-b9w4019h5g | |
tetrapotassium;phosphonato phosphate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS Signal Word: Warning
EINECSNumber : 230-785-7
RUO – Research Use Only